Xylenes, 99+%, for residue analysis, mixed, electronic use grade
Xylenes, 99+%, for residue analysis, mixed, electronic use grade
Xylenes, 99+%, for residue analysis, mixed, electronic use grade
Xylenes, 99+%, for residue analysis, mixed, electronic use grade
Thermo Scientific Chemicals
Xylenes, 99+%, for residue analysis, mixed, electronic use grade
CAS: 1330-20-7 | C8H10
Have Questions?
Catalog NumberQuantityPackaging
FSA3279500101 LGlass Bottle
Catalog number FSA327950010
Price (MYR)
432.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
432.00
Add to cart
Specifications
Chemical Name or MaterialXylenes
Melting Point-34.0°C
Name NoteResidue Free
CAS1330-20-7
Health Hazard 1GHS Signal Word: Warning
View more
For residue analysis
RUO – Research Use Only