CAPSO, 99%
CAPSO, 99%
CAPSO, 99%
Thermo Scientific Chemicals
CAPSO, 99%
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA327741000
Price (MYR)
453.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
453.00
Add to cart
Chemical Identifiers
CAS87-72-9
IUPAC Name(3S,4R,5R)-oxane-2,3,4,5-tetrol
Molecular FormulaC5H10O5
InChI KeySRBFZHDQGSBBOR-ZRMNMSDTSA-N
SMILESO[C@@H]1COC(O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Specific optical rotation+102° to +106° (20°C, 589 nm) (c=10, water)
Residue after ign. (sulfated)=<0.3 %
Heavy metals (as Pb)=<10 ppm
HPLC>=98.5 %
View more
Biological buffer
RUO – Research Use Only