Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals
Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals
Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals
Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals
CAS: 2446-83-5 | C8H14N2O4 | 202.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number FSA327560250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS33228-45-4
IUPAC Name4-hexylaniline
Molecular FormulaC12H19N
InChI KeyOVEMTTZEBOCJDV-UHFFFAOYSA-N
SMILESCCCCCCC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormLiquid
Appearance (Color)Clear colorless to pale yellow to yellow to orange
Identification (FTIR)Conforms
Refractive Index1.5225-1.5265 @ 20?C
C-X Bond Formation (Halogen), Radical initiators
RUO – Research Use Only