Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
Thermo Scientific Chemicals
Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
CAS: 610769-35-2 | C14H30BN3O14 | 475.211 g/mol
Have Questions?
Catalog NumberQuantity
FSA3273300101 L
Catalog number FSA327330010
Price (MYR)
405.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
405.00
Add to cart
Chemical Identifiers
CAS543-49-7
IUPAC Nameheptan-2-ol
Molecular FormulaC7H16O
InChI KeyCETWDUZRCINIHU-UHFFFAOYNA-N
SMILESCCCCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=99.0 %
Specific gravity(20°C) 0.816 to 0.819
Appearance (Form)Liquid
Refractive index1.4200 to 1.4220 (20°C, 589 nm)
View more
Chelation/Complexation reagent, pH buffer
RUO – Research Use Only