Acetic acid, sodium salt, 99+%, for mol biology, anhydrous, DNAse, RNAse and P, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for mol biology, anhydrous, DNAse, RNAse and P, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for mol biology, anhydrous, DNAse, RNAse and P, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for mol biology, anhydrous, DNAse, RNAse and P, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for mol biology, anhydrous, DNAse, RNAse and P, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number FSA327292500
Price (MYR)
553.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
553.00
Add to cart
Chemical Identifiers
CAS462-94-2
IUPAC Namepentane-1,5-diamine
Molecular FormulaC5H14N2
InChI KeyVHRGRCVQAFMJIZ-UHFFFAOYSA-N
SMILESNCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Refractive index1.4570 to 1.4600 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Biological buffer
RUO – Research Use Only