Toluene, 99.85%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number FSA326981000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS75-75-2
IUPAC Namemethanesulfonic acid
Molecular FormulaCH4O3S
InChI KeyAFVFQIVMOAPDHO-UHFFFAOYSA-N
SMILESCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<150 APHA
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Form)Clear liquid
Refractive index1.4285 to 1.4315 (20°C, 589 nm)