1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number FSA326931000
Price (MYR)
438.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
438.00
Add to cart
Chemical Identifiers
CAS7790-30-9
IUPAC Nameλ¹-thallanylium iodide
Molecular FormulaITl
InChI KeyCMJCEVKJYRZMIA-UHFFFAOYSA-M
SMILES[I-].[Tl+]
View more
SpecificationsSpecification SheetSpecification Sheet
FormBeads
Total Metal Impurities0.001% max.
Residual water< 100 ppm
Particle Size-10 Mesh
Appearance (Color)Yellow
Suitable for battery materials development.
RUO – Research Use Only