1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number FSA326930025
Price (MYR)
2,187.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,187.00
Add to cart
Chemical Identifiers
CAS28928-97-4
IUPAC Name3-[(1E)-non-1-en-1-yl]oxolane-2,5-dione
Molecular FormulaC13H20O3
InChI KeyFGQUIQAGZLBOGL-CMDGGOBGNA-N
SMILESCCCCCCCC=CC1CC(=O)OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Refractive Index1.4715-1.4775 @20?C
Appearance (Color)Clear colorless to pale brown
FormLiquid
Suitable for battery materials development.
RUO – Research Use Only