1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
2.5 L
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number FSA326930010
Price (MYR)
1,069.00
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
1,069.00
Add to cart
Specifications
Chemical Name or Material1-Methyl-2-pyrrolidinone
Melting Point-24.0°C
CAS872-50-4
CAS Min %99.4
ColorColorless
View more
Suitable for battery materials development.
RUO – Research Use Only