n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number FSA326920025
Price (MYR)
1,309.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,309.00
Add to cart
Chemical Identifiers
CAS75-52-5
IUPAC Namenitromethane
Molecular FormulaCH3NO2
InChI KeyLYGJENNIWJXYER-UHFFFAOYSA-N
SMILESC[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Lead (Pb)=<0.1 ppm
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %