n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number FSA326920010
Price (MYR)
632.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
632.00
Add to cart
Chemical Identifiers
CAS54451-24-0
IUPAC Namedilanthanum(3+) tricarbonate
Molecular FormulaC3La2O9
InChI KeyNZPIUJUFIFZSPW-UHFFFAOYSA-H
SMILES[La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.