Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.9%, CH2Cl2, CAS Number-75-09-2, 513-35-9, aerothene mm, methylene chloride, freon 30, methylene bichloride, methane, dichloro, methane dichloride, narkotil, methylene dichloride, solmethine, solaesthin, 2.5L, 01, 60, 39.0 deg.C to 40.0 deg.C, CHEBI:15767, Colorless to Yellow
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Catalog number FSA326850025
Price (MYR)
910.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
910.00
Add to cart
Chemical Identifiers
CAS1464-42-2
IUPAC Name2-amino-4-(methylselanyl)butanoic acid
Molecular FormulaC5H11NO2Se
InChI KeyRJFAYQIBOAGBLC-UHFFFAOYNA-N
SMILESC[Se]CCC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-0.5° to +0.5° (20°C, 589 nm) (c=1, 1 N HCl)
Infrared spectrumConforms
Appearance (Form)Glistening powder or flakes
HPLC>=99.0 %
Appearance (Color)Colorless to white to off-white