Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA326820010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS127-47-9
IUPAC Name(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate
Molecular FormulaC22H32O2
InChI KeyQGNJRVVDBSJHIZ-QHLGVNSISA-N
SMILESCC(=O)OCC=C(/C)C=CC=C(/C)C=CC1=C(C)CCCC1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or granules
Assay (unspecified)≥500,000 I.U/g
Appearance (Color)White to cream to yellow to brown
Loss on Drying≤8.0%