Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methylkyanid, methane, cyano, methyl cyanide, mecn, methanecarbonitrile, ethanenitrile, ethyl nitrile, cyanure de methyl, cyanomethane, acetonitrile, 100mL, 0.002 % max., Vapor Pressure: 97mbar at 20 deg.C, 99.9+%, 02, 183, 81.0 deg.C to 82.0 deg.C
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Catalog number FSA326810010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS7782-68-5
IUPAC Namelithium(1+) iodate
Molecular FormulaILiO3
InChI KeyFZAXZVHFYFGNBX-UHFFFAOYSA-M
SMILES[Li+].[O-][I](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder, crystals and chunks
Titration Iodometric>=99.5 %
Heavy metals (as Pb)=<10 ppm
Insoluble matter=<0.01 %