Petroleum ether, for residue analysis, 40° to 60°C, for trace anal. of polyarom. hydrocarbons, Thermo Scientific Chemicals
Petroleum ether, for residue analysis, 40° to 60°C, for trace anal. of polyarom. hydrocarbons, Thermo Scientific Chemicals
Petroleum ether, for residue analysis, 40° to 60°C, for trace anal. of polyarom. hydrocarbons, Thermo Scientific Chemicals
Petroleum ether, for residue analysis, 40° to 60°C, for trace anal. of polyarom. hydrocarbons, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Petroleum ether, for residue analysis, 40° to 60°C, for trace anal. of polyarom. hydrocarbons, Thermo Scientific Chemicals
CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA326790010
Price (MYR)
340.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
340.00
Add to cart
Chemical Identifiers
CAS628-63-7
IUPAC Namepentyl acetate
Molecular FormulaC7H14O2
InChI KeyPGMYKACGEOXYJE-UHFFFAOYSA-N
SMILESCCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4010 to 1.4030 (20°C, 589 nm)
For residue analysis
RUO – Research Use Only