Ethyl acetate, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
Ethyl acetate, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
Ethyl acetate, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
Ethyl acetate, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acetate, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA326610010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl acetate
Melting Point-83.5°C
Name NoteECD Tested, For Pesticide Analysis
CAS141-78-6
ColorColorless
View more
For residue analysis
RUO – Research Use Only