1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals
1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals
1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals
1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals
CAS: 59572-10-0 | C16H6Na4O12S4 | 610.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA325560010
Price (MYR)
517.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
517.00
Add to cart
Chemical Identifiers
CAS71989-26-9
IUPAC Name(2S)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Molecular FormulaC26H32N2O6
InChI KeyUMRUUWFGLGNQLI-ANBDAQEENA-N
SMILESCC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min