Urethane, 97%, Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
CAS: 51-79-6 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA325542500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS5500-21-0
IUPAC Namecyclopropanecarbonitrile
Molecular FormulaC4H5N
InChI KeyAUQDITHEDVOTCU-UHFFFAOYSA-N
SMILESN#CC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4195-1.4235 @ 20?C