Urethane, 97%, Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
CAS: 51-79-6 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Packaging:
Plastic bottle
Catalog number FSA325540010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS592-76-7
IUPAC Namehept-1-ene
Molecular FormulaC7H14
InChI KeyZGEGCLOFRBLKSE-UHFFFAOYSA-N
SMILESCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.3975-1.4015 @ 20?C (UK Sourced Material)
CommentMaterial sourced in UK and US