Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D
Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D
CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Catalog NumberQuantity
FSA3253700505 mL
Catalog number FSA325370050
Price (MYR)
320.00
Add to cart
Quantity:
5 mL
Request bulk or custom format
Price (MYR)
320.00
Add to cart
Chemical Identifiers
CAS71-23-8
IUPAC Namepropan-1-ol
Molecular FormulaC3H8O
InChI KeyBDERNNFJNOPAEC-UHFFFAOYSA-N
SMILESCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Copper (Cu)=<0.02 ppm
Methanol=<0.05 % (GC)
2-Propanol=<0.05 % (GC)
Barium (Ba)=<0.1 ppm
Cobalt (Co)=<0.02 ppm
View more
For NMR
RUO – Research Use Only