Methanol-d4, for NMR, 99.6+% atom D, Thermo Scientific Chemicals
Methanol-d4, for NMR, 99.6+% atom D, Thermo Scientific Chemicals
Methanol-d4, for NMR, 99.6+% atom D, Thermo Scientific Chemicals
Methanol-d4, for NMR, 99.6+% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol-d4, for NMR, 99.6+% atom D, Thermo Scientific Chemicals
Methanol-d4, CH4O, CAS Number-811-98-3, 2h4 methanol, perdeuteromethanol, methan-d3-ol-d, cd3od, trideuterio deuteriooxy methane, methyl-d3 alcohol-d, tetradeuteromethanol, methyl alcohol-d4, methanol-d4, methanol-d4, 99.8 atom % d, 10mL, 01, III, 1187, 65.0 deg.C, Colorless, 0.8880g/mL, 99.60% min.
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
Packaging:
Glass Bottle
Catalog number FSA325360500
Price (MYR)
2,435.00
Add to cart
Quantity:
50 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,435.00
Add to cart
Chemical Identifiers
CAS3393-45-1
IUPAC Name3,6-dihydro-2H-pyran-2-one
Molecular FormulaC5H6O2
InChI KeyVREFDQNWPNDZEX-UHFFFAOYSA-N
SMILESO=C1CC=CCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Colorless to yellow to orange
GC>=88 %
For NMR
RUO – Research Use Only