Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™
Thermo Scientific Chemicals
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™
CAS: 811-98-3 | CH4O | 36.066 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA325351000100 mLAcroSeal™ Glass Bottle
Catalog number FSA325351000
Price (MYR)
3,765.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
3,765.00
Add to cart
Chemical Identifiers
CAS7778-74-7
IUPAC Namepotassium perchlorate
Molecular FormulaClKO4
InChI KeyYLMGFJXSLBMXHK-UHFFFAOYSA-M
SMILES[K+].[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Purity (DSC)≥98.5% (non-U.S. sourced material)
Assay from Suppliers CofA≥98.5% (U.S. sourced material)
Melting Point (DSC)299.0-306.0°C (non-U.S. sourced material)
For NMR
RUO – Research Use Only