Carbon tetrabromide, 98%, Thermo Scientific Chemicals
Carbon tetrabromide, 98%, Thermo Scientific Chemicals
Carbon tetrabromide, 98%, Thermo Scientific Chemicals
Carbon tetrabromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbon tetrabromide, 98%, Thermo Scientific Chemicals
CAS: 558-13-4 | CBr4 | 331.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass Bottle
Catalog number FSA325150050
Price (MYR)
264.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
264.00
Add to cart
Chemical Identifiers
CAS1435-44-5
IUPAC Name1-chloro-2,4-difluorobenzene
Molecular FormulaC6H3ClF2
InChI KeyAJCSNHQKXUSMMY-UHFFFAOYSA-N
SMILESFC1=CC=C(Cl)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4725-1.4765 @ 20?C
C-X bond formation reagent
RUO – Research Use Only