Dimethyl phosphate, 98%
Dimethyl phosphate, 98%
Dimethyl phosphate, 98%
Dimethyl phosphate, 98%
Thermo Scientific Chemicals
Dimethyl phosphate, 98%
CAS: 813-78-5 | C2H7O4P | 126.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3251300101 gGlass bottle
Catalog number FSA325130010
Price (MYR)
763.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
763.00
Add to cart
Specifications
Chemical Name or MaterialDimethyl phosphate
Name Note98%
CAS813-78-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: