(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
CAS: 1139-30-6 | C15H24O | 220.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA325080100
Price (MYR)
312.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
312.00
Add to cart
Chemical Identifiers
CAS97396-56-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Formbeads
CommentSurface area: >550 m2/g
CommentUniformity coefficient: 0.00 - 1.80
Harmonic mean size0.350 - 0.600 mm