Ethylbenzene-d10, for NMR, packaged in ampoules, 99+ atom% D
Ethylbenzene-d10, for NMR, packaged in ampoules, 99+ atom% D
Ethylbenzene-d10, for NMR, packaged in ampoules, 99+ atom% D
Ethylbenzene-d10, for NMR, packaged in ampoules, 99+ atom% D
Thermo Scientific Chemicals
Ethylbenzene-d10, for NMR, packaged in ampoules, 99+ atom% D
CAS: 25837-05-2 | C8H10 | 116.229 g/mol
Have Questions?
Catalog NumberQuantity
FSA3213600101 mL
Catalog number FSA321360010
Price (MYR)
435.00
Add to cart
Quantity:
1 mL
Request bulk or custom format
Price (MYR)
435.00
Add to cart
Chemical Identifiers
CAS150-61-8
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Water Content (Karl Fischer Titration)≤3%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)64.0-70.0°C
Appearance (Color)White to cream or pale pink
View more
For NMR
RUO – Research Use Only