Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Catalog NumberQuantity
FSA3207700757.5 mL
Catalog number FSA320770075
Price (MYR)
374.00
Add to cart
Quantity:
7.5 mL
Request bulk or custom format
Price (MYR)
374.00
Add to cart
Chemical Identifiers
CAS57-09-0
IUPAC Namehexadecyltrimethylazanium bromide
Molecular FormulaC19H42BrN
InChI KeyLZZYPRNAOMGNLH-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with HClO4>=99.0 %
Appearance (Color)White
For NMR
RUO – Research Use Only