Dichloromethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Dichloromethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Dichloromethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Dichloromethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Thermo Scientific Chemicals
Dichloromethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
CAS: 1665-00-5 | CH2Cl2 | 86.939 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3207200757.5 mLAmpule
Catalog number FSA320720075
Price (MYR)
905.00
Add to cart
Quantity:
7.5 mL
Packaging:
Ampule
Request bulk or custom format
Price (MYR)
905.00
Add to cart
Chemical Identifiers
CAS30748-47-1
IUPAC Name1-(2-amino-4-methyl-1,3-thiazol-5-yl)ethan-1-one
Molecular FormulaC6H8N2OS
InChI KeyPKUKCASRNJIQNU-UHFFFAOYSA-N
SMILESCC(=O)C1=C(C)N=C(N)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream or pale yellow to yellow
Assay (HPLC)≥97.0%
FormPowder
Assay (Non-aqueous acid-base Titration)≥97.0 to ≤103.0%
For NMR
RUO – Research Use Only