Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Thermo Scientific Chemicals
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
CAS: 2206-26-0 | C2H3N | 44.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3206600757.5 mLAmpule
Catalog number FSA320660075
Price (MYR)
509.00
Add to cart
Quantity:
7.5 mL
Packaging:
Ampule
Request bulk or custom format
Price (MYR)
509.00
Add to cart
Chemical Identifiers
CAS142-71-2
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with Na2S2O3>=98.5 %
Appearance (Color)Green to blue
Appearance (Form)Crystalline powder
For NMR
RUO – Research Use Only