Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Thermo Scientific Chemicals
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
CAS: 2206-26-0 | C2H3N | 44.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3206500757.5 mLAmpule
Catalog number FSA320650075
Price (MYR)
1,065.00
Add to cart
Quantity:
7.5 mL
Packaging:
Ampule
Request bulk or custom format
Price (MYR)
1,065.00
Add to cart
Chemical Identifiers
CAS7440-23-5
IUPAC Namesodium
Molecular FormulaNa
InChI KeyKEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES[Na]
View more
SpecificationsSpecification SheetSpecification Sheet
FormDispersion in oil
Appearance (Color)Grey
Assay from Supplier's CofASodium: 39.9-40.1%
For NMR
RUO – Research Use Only