Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
CAS: 51364-51-3 | C51H42O3Pd2 | 915.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 mg
Packaging:
Glass bottle
Catalog number FSA318770050
Price (MYR)
2,049.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,049.00
Add to cart
Chemical Identifiers
CAS1422-53-3
IUPAC Name2-bromo-4-fluoro-1-methylbenzene
Molecular FormulaC7H6BrF
InChI KeySFGFOJPGCOYQJK-UHFFFAOYSA-N
SMILESCC1=CC=C(F)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)Clear colorless
Refractive Index1.5265-1.5305 @ 20?C