Silver, 99.9%, (trace metal basis), wire, 0.25 mm diam., 1ea = 10.4gr
Silver, 99.9%, (trace metal basis), wire, 0.25 mm diam., 1ea = 10.4gr
Silver, 99.9%, (trace metal basis), wire, 0.25 mm diam., 1ea = 10.4gr
Silver, 99.9%, (trace metal basis), wire, 0.25 mm diam., 1ea = 10.4gr
Thermo Scientific Chemicals
Silver, 99.9%, (trace metal basis), wire, 0.25 mm diam., 1ea = 10.4gr
CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
FSA3177300101 Ea.
Catalog number FSA317730010
Price (MYR)
1,459.00
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (MYR)
1,459.00
Add to cart
Chemical Identifiers
CAS701-34-8
IUPAC Name4-bromobenzene-1-sulfonamide
Molecular FormulaC6H6BrNO2S
InChI KeySTYQHICBPYRHQK-UHFFFAOYSA-N
SMILESNS(=O)(=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (HPLC)≥97.5%
Melting Point (clear melt)160-172?C
FormPowder
For ultra trace analysis
RUO – Research Use Only