Manganese, 99+%, powder, -40 mesh, Thermo Scientific Chemicals
Manganese, 99+%, powder, -40 mesh, Thermo Scientific Chemicals
Manganese, 99+%, powder, -40 mesh, Thermo Scientific Chemicals
Manganese, 99+%, powder, -40 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Manganese, 99+%, powder, -40 mesh, Thermo Scientific Chemicals
CAS: 7439-96-5 | Mn | 54.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA317442500
Price (MYR)
390.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
390.00
Add to cart
Chemical Identifiers
CAS3087-36-3
IUPAC Nametitanium(4+) tetraethanolate
Molecular FormulaC8H20O4Ti
InChI KeyJMXKSZRRTHPKDL-UHFFFAOYSA-N
SMILES[Ti+4].CC[O-].CC[O-].CC[O-].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Titration Complexometric33 to 35 % (TiO2)
Appearance (Form)Liquid
Refractive index1.5030 to 1.5070 (20°C, 589 nm)