Perrhenic(VII)acid, 99.99%, (trace metal basis), 76.5% solution in water, Thermo Scientific Chemicals
Perrhenic(VII)acid, 99.99%, (trace metal basis), 76.5% solution in water, Thermo Scientific Chemicals
Perrhenic(VII)acid, 99.99%, (trace metal basis), 76.5% solution in water, Thermo Scientific Chemicals
Perrhenic(VII)acid, 99.99%, (trace metal basis), 76.5% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perrhenic(VII)acid, 99.99%, (trace metal basis), 76.5% solution in water, Thermo Scientific Chemicals
CAS: 13768-11-1 | HO4Re | 251.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA316180250
Price (MYR)
3,937.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,937.00
Add to cart
Chemical Identifiers
CAS59662-48-5
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (Silylated GC)≥98.5%
Melting Point (clear melt)218.0-224.0?C
Appearance (Color)White
For ultra trace analysis, Reduction reagent
RUO – Research Use Only