Phosphorus pentoxide, 98+%, ACS reagent, Thermo Scientific Chemicals
Phosphorus pentoxide, 98+%, ACS reagent, Thermo Scientific Chemicals
Phosphorus pentoxide, 98+%, ACS reagent, Thermo Scientific Chemicals
Phosphorus pentoxide, 98+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorus pentoxide, 98+%, ACS reagent, Thermo Scientific Chemicals
CAS: 1314-56-3 | O5P2 | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number FSA315820010
Price (MYR)
940.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
940.00
Add to cart
Chemical Identifiers
CAS939-26-4
IUPAC Name2-(bromomethyl)naphthalene
Molecular FormulaC11H9Br
InChI KeyRUHJZSZTSCSTCC-UHFFFAOYSA-N
SMILESBrCC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light beige
Infrared spectrumConforms
GC>=95 %
Appearance (Form)Crystalline powder
Melting point52°C to 57°C
Adsorbent
RUO – Research Use Only