Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA315180025
Price (MYR)
3,436.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
3,436.00
Add to cart
Chemical Identifiers
CAS2855-13-2
IUPAC Name3-(aminomethyl)-3,5,5-trimethylcyclohexan-1-amine
Molecular FormulaC10H22N2
InChI KeyRNLHGQLZWXBQNY-UHFFFAOYNA-N
SMILESCC1(C)CC(N)CC(C)(CN)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Clear liquid
Refractive index1.4880 to 1.4900 (20°C, 589 nm)
GC>=99.0 % (Sum of isomers)
Water=<0.5 % (K.F.)
View more
For ultra trace analysis, Lewis acid
RUO – Research Use Only