Check Order Status
3-Chloropropyl isocyanate, 96%, Thermo Scientific™
Element Search
Structure Search
3-Chloropropyl isocyanate, 96%, Thermo Scientific™
Have Questions?
Change view
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number
FSA314590010
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
3-Chloropropyl isocyanate
Name Note
96%
CAS
13010-19-0
CAS Min %
95.0
Health Hazard 1
GHS Signal Word:
Danger