Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
CAS: 4461-30-7 | C3H2ClNO2 | 119.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA314300050
Price (MYR)
238.51
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
238.51
Add to cart
Chemical Identifiers
CAS105-87-3
IUPAC Name(2E)-3,7-dimethylocta-2,6-dien-1-yl acetate
Molecular FormulaC12H20O2
InChI KeyHIGQPQRQIQDZMP-DHZHZOJOSA-N
SMILESCC(C)=CCC\C(C)=C\COC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Refractive Index1.4595-1.4635 @ 20?C