Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
CAS: 4461-30-7 | C3H2ClNO2 | 119.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA314300010
Price (MYR)
83.52
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
83.52
Add to cart
Specifications
Chemical Name or MaterialChloroacetyl isocyanate
Name Notetech., 90%
CAS4461-30-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.