1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
CAS: 2708-97-6 | C6F4I2 | 401.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA311690010
Price (MYR)
551.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
551.00
Add to cart
Chemical Identifiers
CAS12064-62-9
IUPAC Namedigadolinium(3+) trioxidandiide
Molecular FormulaGd2O3
InChI KeyCMIHHWBVHJVIGI-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Gd+3].[Gd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total conc. of imp.as REO=<0.1 %
Appearance (Form)Powder