Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number FSA307050100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS78-83-1
IUPAC Name2-methylpropan-1-ol
Molecular FormulaC4H10O
InChI KeyZXEKIIBDNHEJCQ-UHFFFAOYSA-N
SMILESCC(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Water=<0.005 %
Residue after evaporation=<0.001 %