Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals
Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals
Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals
Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals
CAS: 5188-07-8 | CH3NaS | 70.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA305710250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS77-86-1
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC4H11NO3
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
CommentComponents are dissolved in UV treated 18.2 megohm-cm ± 1 water and filtered through 1 micron filter.
FormLiquid
pH7.4 ± 0.15