4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
CAS: 453-71-4 | C7H4FNO4 | 185.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA305060250
Price (MYR)
1,210.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,210.00
Add to cart
Chemical Identifiers
CAS148980-31-8
IUPAC Nameterbium(3+) tritrifluoromethanesulfonate
Molecular FormulaC3F9O9S3Tb
InChI KeyJLYAVYSVLLIWEK-UHFFFAOYSA-K
SMILES[Tb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (unspecified)Terbium: ≥25.6% (theoretical: 26.2%)