4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
CAS: 453-71-4 | C7H4FNO4 | 185.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA305060050
Price (MYR)
388.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
388.00
Add to cart
Chemical Identifiers
CAS6759-78-0
IUPAC Name8-hydroxyquinoline-2-carbonitrile
Molecular FormulaC10H6N2O
InChI KeyKUQKKIBQVSFDHX-UHFFFAOYSA-N
SMILESOC1=C2N=C(C=CC2=CC=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=97.5 %
Melting point131°C to 136°C