4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals
4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals
4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals
4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals
CAS: 1120-97-4 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA305000050
Price (MYR)
272.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
272.00
Add to cart
Chemical Identifiers
CAS7647-10-1
IUPAC Namepalladium(2+) dichloride
Molecular FormulaCl2Pd
InChI KeyPIBWKRNGBLPSSY-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Copper (Cu)=<15 ppm
Magnesium (Mg)=<15 ppm
Silicon (Si)=<30 ppm
Appearance (Color)Red-brown
Assay59 to 60 % (Pd)