Harmine, 98%, Thermo Scientific Chemicals
Harmine, 98%, Thermo Scientific Chemicals
Harmine, 98%, Thermo Scientific Chemicals
Harmine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Harmine, 98%, Thermo Scientific Chemicals
CAS: 442-51-3 | C13H12N2O | 212.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
Catalog number FSA302972500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS13395-16-9
IUPAC Namecopper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14CuO4
InChI KeyQYJPSWYYEKYVEJ-FDGPNNRMSA-L
SMILES[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Blue or blue-grey to purple
FormPowder
Assay (Redox Titration)≥97.5 to ≤102.5%