Linolenic acid, 99%
Linolenic acid, 99%
Linolenic acid, 99%
Thermo Scientific Chemicals
Linolenic acid, 99%
CAS: 463-40-1 | C18H30O2 | 278.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA302825000
Price (MYR)
618.00
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
618.00
Add to cart
Chemical Identifiers
CAS10294-41-4
IUPAC Namecerium(3+) hexahydrate trinitrate
Molecular FormulaCeH12N3O15
InChI KeyQQZMWMKOWKGPQY-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline Aggregates
Assay (special)38.8-40.5% CeO₂
Total Rare Earth Oxide Impurities≤0.01% (100 ppm)