(+)-Dibenzoyl-D-tartaric acid, 98+%, Thermo Scientific Chemicals
(+)-Dibenzoyl-D-tartaric acid, 98+%, Thermo Scientific Chemicals
(+)-Dibenzoyl-D-tartaric acid, 98+%, Thermo Scientific Chemicals
(+)-Dibenzoyl-D-tartaric acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(+)-Dibenzoyl-D-tartaric acid, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA302515000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS25322-68-3
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
CompositionLithium Acetate Dihydrate: 100 mM
FormLiquid
CompositionPEG: 50%
Appearance (Color)Clear, colorless