2-Chloroethylphosphonic acid, 90%, Tech., Thermo Scientific Chemicals
2-Chloroethylphosphonic acid, 90%, Tech., Thermo Scientific Chemicals
2-Chloroethylphosphonic acid, 90%, Tech., Thermo Scientific Chemicals
2-Chloroethylphosphonic acid, 90%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloroethylphosphonic acid, 90%, Tech., Thermo Scientific Chemicals
CAS: 16672-87-0 | C2H6ClO3P | 144.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA301630010
Price (MYR)
794.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
794.00
Add to cart
Chemical Identifiers
CAS13780-48-8
IUPAC Nameλ²-europium(2+) dibromide
Molecular FormulaBr2Eu
InChI KeyPJVPGMOCWCUQHP-UHFFFAOYSA-L
SMILES[Br-].[Br-].[Eu++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.