1-Amino-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals
1-Amino-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals
1-Amino-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals
1-Amino-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Amino-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals
CAS: 22059-21-8 | C4H7NO2 | 101.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
Glass bottle
Catalog number FSA301420010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7758-98-7
IUPAC Namecopper(2+) sulfate
Molecular FormulaCuO4S
InChI KeyARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES[Cu++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<1 % (300°C)
Total nitrogen (as N)=<0.01 %
Calcium (Ca)=<20 ppm
Magnesium (Mg)=<50 ppm
Appearance (Color)Light green to grey